Learn More
MSC Lubriplate Grease, NLGI 2, Lithium, High Temperature, 14.5 oz
Catalog No. 501112923
Encompass
Lubriplate No. 1200-2 is a general purpose grease designed for heavy-duty applications requiring high film strength, greater stability, and extended lubricant life. It provides a tough protective film for superior lubrication in automotive, construction, and industrial equipment, offering protection against shock loads, mud, and water.
  • Exceptional film strength
  • Excellent shear stability at high temperatures
  • Highly waterproof
  • Anti-corrosion properties
  • Multi-purpose for various heavy-duty applications
Catalog No. 50-111-2923 Supplier MSC Supplier No. 01245422
Lubriplate No. 1200-2 is a general purpose grease designed for heavy-duty applications requiring high film strength, greater stability, and extended lubricant life. It provides a tough protective film for superior lubrication in automotive, construction, and industrial equipment, offering protection against shock loads, mud, and water.
  • Exceptional film strength
  • Excellent shear stability at high temperatures
  • Highly waterproof
  • Anti-corrosion properties
  • Multi-purpose for various heavy-duty applications