Learn More
Sino Biological Recombinant Human Galectin-7/LGALS7 Protein (His Tag)
Supplier:  Sino Biological 12000H07E100
Encompass_Preferred
A DNA sequence encoding the human LGALS7 (NP_002298.1) (Ser2-Phe136) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7712