Learn More
Ace Glass, Inc. Separatory funnel, 12000mL, 55/50, 10mm bore glass stopcock
SDP
Catalog No. 50979213
Encompass
12000mL separatory, globe type funnel, with neck and section between globe and stopcock shaped to provide a secure grip, for shaking contents or transporting. Funnel features a 55/50 standard taper top joint, and a 10mm bore glass stopcock plug.
Catalog No. 50-979-213 Supplier Ace Glass, Inc. Supplier No. 722433
Add to Cart
Add to Cart
12000mL separatory, globe type funnel, with neck and section between globe and stopcock shaped to provide a secure grip, for shaking contents or transporting. Funnel features a 55/50 standard taper top joint, and a 10mm bore glass stopcock plug.