Learn More
Ace Glass, Inc. 12000mL hemispherical fabric mantle, Glas-Col, 293mm max flask diameter, 6.27in inside depth, 14.53in OD, 7.77in height, 2 circuits at 650 watts each, CSA rated
SDP
Catalog No. 50977269
Encompass
12000mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-977-269 Supplier Ace Glass, Inc. Supplier No. 1203527
12000mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.