Learn More
Chemglass Life Sciences Glas-col Mantle Tripod Support for Series inMin, Mantle Size 12000mL, 14in Fixed Height, 425mm ID
SDP
Catalog No. CG1100603
Encompass_Preferred
Glas-col Mantle Tripod Support for Series inMin, Mantle Size 12000mL, 14in Fixed Height, 425mm ID. Glas-Col tripod support sits on the floor or a bench to hold a Series M mantle ranging in sizes from 3 to 22 liters. Mounting holes are in the foot of each leg for securing to floor or bench. Support is painted gray and shipped unassembled.
Catalog No. CG1100603 Supplier Chemglass Life Sciences Supplier No. CG1100603
Glas-col Mantle Tripod Support for Series inMin, Mantle Size 12000mL, 14in Fixed Height, 425mm ID. Glas-Col tripod support sits on the floor or a bench to hold a Series M mantle ranging in sizes from 3 to 22 liters. Mounting holes are in the foot of each leg for securing to floor or bench. Support is painted gray and shipped unassembled.