Learn More
ADMINMED NANOBIOSCIENCES 1200 Microneedle Liquid Injection System
SDP
Catalog No. NC2292701
Encompass
AdminPen 1200 microneedle liquid injection system is suitable for attachment to a standard syringe. The system has forty three 1100 um-tall microneedles on 1cm2 circular microneedle array. The microneedle array is made of SS316L stainless steel.
This item is not returnable. View return policy
Catalog No. NC2292701 Supplier ADMINMED NANOBIOSCIENCES Supplier No. 402
AdminPen 1200 microneedle liquid injection system is suitable for attachment to a standard syringe. The system has forty three 1100 um-tall microneedles on 1cm2 circular microneedle array. The microneedle array is made of SS316L stainless steel.