Learn More
Chemglass Life Sciences Dish, Crystallizing, 1200mL, 150 x 75mm
SDP
Catalog No. 501215012
Encompass_Preferred
Dish, Crystallizing, 1200mL, 150 x 75mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.
Catalog No. 50-121-5012 Supplier Chemglass Life Sciences Supplier No. CG8276P150
Dish, Crystallizing, 1200mL, 150 x 75mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.