Learn More
ADMINMED NANOBIOSCIENCES 1200UM MICRONEEDLE ARRAY
SDP
Catalog No. NC0584389
Encompass
AdminPatch Array 1200 is a microneedle array, which has 43 (forty three) 1100 um-tall microneedles located within 1 sq. cm circular area. The entire device is 20 mm in diameter and is made of medical-grade SS316L stainless steel.
Catalog No. NC0584389 Supplier ADMINMED NANOBIOSCIENCES Supplier No. SKU102
AdminPatch Array 1200 is a microneedle array, which has 43 (forty three) 1100 um-tall microneedles located within 1 sq. cm circular area. The entire device is 20 mm in diameter and is made of medical-grade SS316L stainless steel.