Learn More
Sino Biological Recombinant Human Vitamin D Receptor/VDR Protein (His Tag)
Supplier:  Sino Biological 12025H08B50
Encompass_Preferred
A DNA sequence encoding the human VDR (P11473) (Met 1-Ser 427) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7725