Learn More
Sino Biological Recombinant Human HEXA Protein (Subunit A, His Tag)
Supplier:  Sino Biological 12037H08B50
Encompass_Preferred
A DNA sequence encoding the human HEXA (AAD13932.1) (Met 1-Thr 529) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7731