Learn More
Sino Biological Recombinant Human IL-17A Protein (T26A, His Tag)
Supplier:  Sino Biological 12047H07B100
Encompass_Preferred
A DNA sequence encoding the mature form of human IL17A (Q16552, with mutation Thr 26 Ala) (Gly24-Ala155) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7748