Learn More
Sino Biological Recombinant Human IL-17A Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 12047HNAEB20
Encompass_Preferred
A DNA sequence encoding the human IL17A (Q16552) (Ile20-Ala155) was expressed, with a N-terminal Met. The purified protein was biotinylated in vitro.
Catalog No. 50-161-7757