Learn More
Sino Biological Recombinant Human GGH/Glutamyl hydrolase gamma Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12068H08H100
Encompass_Preferred
A DNA sequence encoding the human GGH (NP_003869.1) (Met1-Asp318) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7769