Learn More
Sino Biological Recombinant Human NRAS Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12073H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human NRAS (P01111) (Met1-Cys186) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7779