Learn More
Sino Biological Recombinant Human GLA/alpha-Galactosidase A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12078H08H50
Encompass_Preferred
A DNA sequence encoding the human GLA (NP_000160.1) (Met 1-Leu 429) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7784