Learn More
Sino Biological Recombinant Human Transthyretin Protein (His Tag)
Supplier:  Sino Biological 12091H07H100
Encompass_Preferred
A DNA sequence encoding the human TTR (NP_000362.1) (Gly21-Glu147) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7797