Learn More
Sino Biological Recombinant Human DR3/TNFRSF25 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 12095H02H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF25 (NP_683867.1) (Met1-Gln199) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7803