Learn More
MSC Securall Cabinets Vertical Drum Cabinet, 120 gal., 1 ea
Catalog No. 502886343
Encompass
This vertical drum cabinet from Securall Cabinets is constructed from 18 gauge steel and measures 56 in wide x 31 in deep x 65 in high. It is designed to safely store two drums.
  • Features a 3 point key lock for security
  • Yellow finish
  • Manual closing door mechanism
  • Includes one shelf
  • Designed to accommodate two drums
  • Drum rollers included for ease of use
Catalog No. 50-288-6343 Supplier MSC Supplier No. 76921733
This vertical drum cabinet from Securall Cabinets is constructed from 18 gauge steel and measures 56 in wide x 31 in deep x 65 in high. It is designed to safely store two drums.
  • Features a 3 point key lock for security
  • Yellow finish
  • Manual closing door mechanism
  • Includes one shelf
  • Designed to accommodate two drums
  • Drum rollers included for ease of use