Learn More
MOLDPRO INC 120 mL Titration Beaker, 42.3 x 113 mm, Clear, 300/case, Flat Bottom, PP Copolymer
Catalog No. NC1779905
Encompass_Preferred
MoldPro MP-117 120 mL Titration Beaker used in wet chemistry titration applications especially TAN and TBN. Direct replacement for Metrohm 6.1459.300 and 6.1459.307 absent threads and threaded cap option. Fits multiple USB and Robotic Sample platforms used with Metrohm Titration systems. • 300 beakers/case • Made from virgin polypropylene • Excellent visibility • Good resistance to acids Fit Metrohm sample introduction systems: • 814 USB Sample Processor • 815 Robotic USB Sample Processor XL • 815 Robotic Soliprep • 855 Robotic Titrosampler • 864 Robotic Balance Sample Processor • 898 XYZ Sample Changer • MATi 02 Fit Metrohm racks: • 6.2041.470 • 6.2041.840 • 6.2041.850 • 6.2068.010 • 6.2068.020 • 6.2071.020 • 6.2071.030 • 6.9920.141 Methods: • ASTM D 664 • ASTM D 2896
Catalog No. NC1779905 Supplier MOLDPRO INC Supplier No. MP117
MoldPro MP-117 120 mL Titration Beaker used in wet chemistry titration applications especially TAN and TBN. Direct replacement for Metrohm 6.1459.300 and 6.1459.307 absent threads and threaded cap option. Fits multiple USB and Robotic Sample platforms used with Metrohm Titration systems. • 300 beakers/case • Made from virgin polypropylene • Excellent visibility • Good resistance to acids Fit Metrohm sample introduction systems: • 814 USB Sample Processor • 815 Robotic USB Sample Processor XL • 815 Robotic Soliprep • 855 Robotic Titrosampler • 864 Robotic Balance Sample Processor • 898 XYZ Sample Changer • MATi 02 Fit Metrohm racks: • 6.2041.470 • 6.2041.840 • 6.2041.850 • 6.2068.010 • 6.2068.020 • 6.2071.020 • 6.2071.030 • 6.9920.141 Methods: • ASTM D 664 • ASTM D 2896