Learn More
Sarstedt Inc Tube 120ml, 114x44mm, PP
Supplier:  Sarstedt Inc 60.597.001
Encompass_Preferred
Screw cap tube, 120 ml, 114 x 44 mm,flat base, PP, with assembled cap, sterile, 25 pieces per bag
Catalog No. NC9655914