Learn More
Chemglass Life Sciences Reaction Kettle Lid, Duran, Flat Ground, Lid ID: 120mm
SDP
Catalog No. CG14903
Encompass_Preferred
Reaction Kettle Lid, Duran, Flat Ground, Lid ID: 120mm. Dome style lid for construction of single or multiple neck reaction kettle heads. Mating surface is ground. Outer taper of flange is formed at a 45 angle to accept the CG-141 stainless steel clamps.
Catalog No. CG14903 Supplier Chemglass Life Sciences Supplier No. CG14903
Reaction Kettle Lid, Duran, Flat Ground, Lid ID: 120mm. Dome style lid for construction of single or multiple neck reaction kettle heads. Mating surface is ground. Outer taper of flange is formed at a 45 angle to accept the CG-141 stainless steel clamps.