Learn More
Chemglass Life Sciences Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-04
SDP
Catalog No. CG159804
Encompass_Preferred
Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-04. Vented polyethylene stopper to minimize evaporation from Dewar flasks.
Catalog No. CG159804 Supplier Chemglass Life Sciences Supplier No. CG159804
Polyethylene Stopper for Dewar Flask, Fits CG-1593 & CG-1594-04. Vented polyethylene stopper to minimize evaporation from Dewar flasks.