Learn More
Glas Col LLC Shaker
SDP
Catalog No. NC9751630
Encompass_Preferred
Bench top shaker base, motion to simulate hand shaking. 2-125 RPM. Variable speed motor for gentle low speed to vigorous high speed shaking. 120V, 1.0A, 50/60Hz. Can be used for up to 20 pound loads.
Catalog No. NC9751630 Supplier Glas Col LLC Supplier No. 099A S60012
Bench top shaker base, motion to simulate hand shaking. 2-125 RPM. Variable speed motor for gentle low speed to vigorous high speed shaking. 120V, 1.0A, 50/60Hz. Can be used for up to 20 pound loads.