Learn More
Thermo Scientific Replacement 120V solenoid valve
Supplier:  Thermo Scientific 4000617
Encompass_Preferred
Replacement 120V solenoid valve for Models 7450 and 7470 Series CryoMed Controlled Rate Freezers.
Catalog No. NC1797884