Learn More
Sino Biological Recombinant Human MUC-1 Protein (mFc Tag)
Supplier:  Sino Biological 12123H05H100
Encompass_Preferred
A DNA sequence encoding the human MUC1 isoform 2 (NP_001018016.1) extracellular domain (Met1-Gly167) was fused with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-7817