Learn More
Sino Biological Recombinant Human MUC-1 Protein
Supplier:  Sino Biological 12123HCCH100
Encompass_Preferred
A DNA sequence encoding the human MUC1 isoform 2 (NP_001018016.1) extracellular domain (Met 1-Gly 167) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-161-7819