Learn More
Sino Biological Recombinant Human PCNA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12131H07B100
Encompass_Preferred
A DNA sequence encoding the mature form of human PCNA (P12004) (Met 1-Ser 261) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7821