Learn More
Sino Biological Recombinant Human SRPK3 Protein (His & GST Tag)
Supplier:  Sino Biological 12132H20B50
Encompass_Preferred
A DNA sequence encoding full length of human SRPK3 isoform 2 (NP_001164231.1) (Met 1-Pro 566) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7824