Learn More
Sino Biological Recombinant Human LSAMP Protein (hFc Tag)
Supplier:  Sino Biological 12136H02H100
Encompass_Preferred
A DNA sequence encoding the human LSAMP (Q13449) (Met 1-Asn 315), without the pro peptide, was was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7825