Learn More
Sino Biological Recombinant Human Carbonic Anhydrase VII Protein (His Tag)
Supplier:  Sino Biological 12147H08E100
Encompass_Preferred
A DNA sequence encoding the human CA7 (P43166) (Met 1-Ala 264) was fused with a polyhistide tag at the C-terminus.
Catalog No. 50-161-7839