Learn More
Sino Biological Recombinant Human Syndecan-3 Protein (ECD, His Tag)
Supplier:  Sino Biological 12158H08H200
Encompass_Preferred
A DNA sequence encoding the human SDC3 (ABM82826.1) (Met1-Lys311) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7843