Learn More
Sino Biological Recombinant Human Tim4/TIMD4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12161H08H100
Encompass_Preferred
A DNA sequence encoding the human TIMD4 (AAH08988.1) extracellular domain (Met 1-Leu 315) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7845