Learn More
Sino Biological Recombinant Human VSIG4 Protein (hFc Tag)
Supplier:  Sino Biological 12163H02H100
Encompass_Preferred
A DNA sequence encoding the human VSIG4 (NP_009199.1) (Met1-Pro283) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7849