Learn More
Sino Biological Recombinant Human VSIG4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12163H08H100
Encompass_Preferred
A DNA sequence encoding the human VSIG4 (Q9Y279-1) extracellular domain (Met 1-Pro 283) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7851