Learn More
Sino Biological Recombinant Human SIGIRR Protein (hFc Tag)
Supplier:  Sino Biological 12165H02H100
Encompass_Preferred
A DNA sequence encoding the human SIGIRR (Q6IA17-1) (Met1-His118) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7853