Learn More
Sino Biological Recombinant Human Alpha-fetoprotein Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12177H08H50
Encompass_Preferred
A DNA sequence encoding the human AFP (P02771) (Met 1-Val 609) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7858