Learn More
Sino Biological Recombinant Human PDZD11 Protein (His Tag)
Supplier:  Sino Biological 12188H07E100
Encompass_Preferred
A DNA sequence encoding the human PDZD11 (NP_057568.1) (Asp 2-His 140) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-7869