Learn More
Sino Biological Recombinant Human CD160 Protein (His Tag)
Supplier:  Sino Biological 12191H08H100
Encompass_Preferred
A DNA sequence encoding the human CD160 (NP_008984.1) (Met 1-Leu 158) without the propeptide was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7871