Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 70% (v/v), Solvent Grade, Ricca Chemical
SDP

Catalog No. R42100704A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
4 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R42100704A 4 L Natural Poly Bottle
R421007020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R42100704A Supplier Ricca Chemical Company Supplier No. R42100704A

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
Appearance Colorless liquid
Quantity 4 L
Packaging Natural Poly Bottle
Specific Gravity At 20°C: 0.874-0.882
Assay Percent Range 68 to 72% v/v
Chemical Name or Material Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.