Learn More
Sino Biological Recombinant Human Betacellulin Protein (hFc Tag)
Catalog No. 501617873
Encompass_Preferred
A DNA sequence encoding the human BTC (P35070) (Met1-Tyr111) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7873 Supplier Sino Biological Supplier No. 12192H02H100
A DNA sequence encoding the human BTC (P35070) (Met1-Tyr111) was expressed with the Fc region of human IgG1 at the C-terminus.