Learn More
Sino Biological Recombinant Human STK16 Protein (His & NusA Tag)
Supplier:  Sino Biological 12201H24E50
Encompass_Preferred
A DNA sequence encoding the native human STK16 (AAH02618.1) (Met 1-Ile 305) was fused with the polyhistidine-tagged NusA tag at the N-terminus.
Catalog No. 50-161-7881