Learn More
Sino Biological Recombinant Human CD44 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 12211H02H100
Encompass_Preferred
A DNA sequence encoding the human CD44 (P16070-1) (Met1-Pro220) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-7884