Learn More
Sino Biological Recombinant Human CD44 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12211H08H100
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Met 1-Pro 220) of human CD44 (P16070-1) extracellular domain was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7886