Learn More
Sino Biological Recombinant Human TLK1 Protein (His & GST Tag)
Supplier:  Sino Biological 12213H20B50
Encompass_Preferred
A DNA sequence encoding the human TLK1 isoform 1 (Q9UKI8-1) (Met 1-Tyr 766) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-7889