Learn More
Sino Biological Recombinant Human COL9A1 Protein (mFc Tag)
Supplier:  Sino Biological 12231H05H100
Encompass_Preferred
A DNA sequence encoding the human COL9A1 (P20849-3) (Met1-Pro328) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-7898