Learn More
Sino Biological Recombinant Human COL9A1 Protein (His Tag)
Supplier:  Sino Biological 12231H08H100
Encompass_Preferred
A DNA sequence encoding the human COL9A1(P20849-3) (Met1-Pro328) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-7900