Learn More
Sino Biological Recombinant Human TH1R/PTHR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12232H08H100
Encompass_Preferred
A DNA sequence encoding the human PTH1R (Q03431) extracellular domain (Met 1-Gly 188) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7902