Learn More
Sino Biological Recombinant Human Siglec-3/CD33 Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 12238H05H100
Encompass_Preferred
A DNA sequence encoding the human CD33 (AAH28152.1) (Met1-His259) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-7904