Learn More
Sino Biological Recombinant Human Siglec-3/CD33 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 12238H08H100
Encompass_Preferred
A DNA sequence encoding the human CD33 (AAH28152.1) (Met1-His259) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-7906