Learn More
Sino Biological Recombinant Human CD46 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12239H08H50
Encompass_Preferred
A DNA sequence encoding the human CD46 (EAW93471.1) extracellular domain (Met 1-Asp 328) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-7912